Avanafil 100 µg/mL in Acetonitrile:Dimethylsulfoxide, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-466
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:330784-47-9
- Description:Avanafil 100 µg/mL in Acetonitrile:Dimethylsulfoxide
- Size:1 ml
- Formula:C₂₃H₂₆ClN₇O₃
- Molecular Weight:483.951 g/mol
- Cat. No.:77691-466