2-Amino-4-methoxy-6-methyl-1,3,5-triazine 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77691-457
New Product
:
- CAS No.:1668-54-8
- Description:2-Amino-4-methoxy-6-methyl-1,3,5-triazine 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₅H₈N₄O
- Molecular Weight:140.1400 g/mol
- Cat. No.:77691-457