Order Entry
Puerto Rico
ContactUsLinkComponent
 
4-Amino-3-fluorophenol 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-456
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
4-Amino-3-fluorophenol 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-456
Supplier:  LGC Standards
Supplier Number:  DRE-A10202400AL-100
New Product
CAS Number:  

Specifications

  • CAS No.:
    399-95-1
  • Description:
    4-Amino-3-fluorophenol 100 µg/mL in Acetonitrile
  • Size:
    1 ml
  • Formula:
    C₆H₆FNO
  • Molecular Weight:
    127.12 g/mol
  • Cat. No.:
    77691-456

Specifications

About this item