Albendazole 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-447
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:54965-21-8
- Description:Albendazole 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₂H₁₅N₃O₂S
- Molecular Weight:265.33 g/mol
- Cat. No.:77691-447