Acid Violet 49 100 µg/mL in Ethanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-443
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:1694-09-3
- Description:Acid Violet 49 100 µg/mL in Ethanol
- Size:1 ml
- Formula:C₃₉H₄₁N₃O₆S₂
- Molecular Weight:733.87 g/mol
- Cat. No.:77691-443