Acetic acid-methyl ester 1000 µg/mL in Methanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-435
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:79-20-9
- Description:Acetic acid-methyl ester 1000 µg/mL in Methanol
- Size:1 ml
- Formula:C₃H₆O₂
- Molecular Weight:74.08 g/mol
- Cat. No.:77691-435