Acetic acid-isobutyl ester 1000 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-434
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:110-19-0
- Description:Acetic acid-isobutyl ester 1000 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₆H₁₂O₂
- Molecular Weight:116.16 g/mol
- Cat. No.:77691-434