4-Acetylaminoantipyrine 10 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77691-430
New Product
:
- CAS No.:83-15-8
- Description:4-Acetylaminoantipyrine 10 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₃H₁₅N₃O₂
- Molecular Weight:245.28 g/mol
- Cat. No.:77691-430