Order Entry
Puerto Rico
ContactUsLinkComponent
2-(4-Aminophenyl)ethylamine ≥97%
Irritant
Corrosive
2-(4-Aminophenyl)ethylamine ≥97%
Catalog # ALA151503-1G
Supplier:  Aladdin Scientific
New Product
CAS Number:  
undefined
Irritant
Corrosive
2-(4-Aminophenyl)ethylamine ≥97%
Catalog # ALA151503-1G
Supplier:  Aladdin Scientific
Supplier Number:  A151503-1G
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Cat. No.:
    ALA151503-1G

Specifications

About this item

 :
 :136.20 g/mol
Boiling Pt:142 °C
Melting Pt:28 °C
Density:1.034 g/cm³
Flash Pt:113 °C
 :GHS07
 :GHS05
Storage Temperature:Ambient
CAS Number:13472-00-9
Formula:C₈H₁₂N₂
MDL Number:MFCD00008194