Order Entry
Puerto Rico
ContactUsLinkComponent
 
25-Azido-2,5,8,11,14,17,20,23-octaoxapentacosane ≥90% (by HPLC)
Catalog # ALA151379-25MG
Supplier:  Aladdin Scientific
New Product
CAS Number:  
undefined
25-Azido-2,5,8,11,14,17,20,23-octaoxapentacosane ≥90% (by HPLC)
Catalog # ALA151379-25MG
Supplier:  Aladdin Scientific
Supplier Number:  A151379-25MG
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Cat. No.:
    ALA151379-25MG

Specifications

About this item

 :
 :409.48 g/mol
Storage Temperature:Refrigerator
CAS Number:869718-80-9
Formula:C₁₇H₃₅N₃O₈
MDL Number:MFCD13184962