Azetidin-2-one ≥97%
About this item
It is an unsubstituted four membered lactam.It is the fundamental building block of β-lactam antibiotics. X-ray photoelectron spectra of 2-azetidinone was investigated.It undergoes hydrolysis with aqueous alkali to yield β-alanine.It was used in synthesis of optically pure densely functionalized γ-lactams.
- CAS Number:930-21-2
- Formula:C₃H₅NO
- MDL Number:MFCD00013328
- Molecular Weight:71.08 g/mol
- Boiling Pt:106 °C
- Melting Pt:74-76°C
- Storage Temperature:Refrigerator
4 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
It is an unsubstituted four membered lactam.It is the fundamental building block of β-lactam antibiotics. X-ray photoelectron spectra of 2-azetidinone was investigated.It undergoes hydrolysis with aqueous alkali to yield β-alanine.It was used in synthesis of optically pure densely functionalized γ-lactams.
- CAS Number:930-21-2
- Formula:C₃H₅NO
- MDL Number:MFCD00013328
- Molecular Weight:71.08 g/mol
- Boiling Pt:106 °C
- Melting Pt:74-76°C
- Storage Temperature:Refrigerator
Product Family Options
Product Information
- AvailabilityPrice
- Supplier Number: A139449-1GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: A139449-5GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: A139449-10GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: A139449-25GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



