Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Azithromycin
Azithromycin, Moligand™ analytical standard
Azithromycin, Moligand™ analytical standard
ALA134451-25MG
:
ALADDIN SCIENTIFIC
New Product
:
Azithromycin, Moligand™ analytical standard
ALA134451-25MG
:
ALADDIN SCIENTIFIC
:
A134451-25MG
New Product
:
Cat. No.:
ALA134451-25MG
:
:
749 g/mol
:
GHS07
:
GHS09
:
GHS08
:
83905-01-5
:
C₃₈H₇₂N₂O₁₂
:
MFCD00873574