Order Entry
Puerto Rico
ContactUsLinkComponent
9-Anthrylmethyl methacrylate ≥98%
Irritant
Corrosive
9-Anthrylmethyl methacrylate ≥98%
Catalog # ALA124641250MG
Supplier:  Aladdin Scientific
New Product
CAS Number:  
undefined
Irritant
Corrosive
9-Anthrylmethyl methacrylate ≥98%
Catalog # ALA124641250MG
Supplier:  Aladdin Scientific
Supplier Number:  A124641-250MG
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Cat. No.:
    ALA124641250MG

Specifications

About this item

 :
 :276.34 g/mol
Melting Pt:80-90°C
 :GHS07
 :GHS05
Storage Temperature:Refrigerator
CAS Number:31645-35-9
Formula:C₁₉H₁₆O₂
MDL Number:MFCD00673247