California Pesticide Class 2A Mixture Kit; Primary and secondary source, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-025
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Acetonitrile:Formic Acid
- Pack type:Glass bottle
- Description:California Pesticide Class 2A Mi xture Kit; Primary and secondary source
- Size:2 x 1 ml
- Cat. No.:77679-025