Order Entry
Puerto Rico
ContactUsLinkComponent
Ammonium metavanadate ≥99% AR
Health
Toxic
Ammonium metavanadate ≥99% AR
Catalog # ALA111822-500G
Supplier:  Aladdin Scientific
New Product
CAS Number:  
Health
Toxic
Ammonium metavanadate ≥99% AR
Catalog # ALA111822-500G
Supplier:  Aladdin Scientific
Supplier Number:  A111822-500G
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

About this item

 :116.98 g/mol
Melting Pt:200°C
Density:2.32 g/cm³
EINECS:232-261-3
UN:2859
ADR:6.1,II
 :GHS08
 :GHS06
CAS Number:7803-55-6
 :MFCD00011453
Formula:NH₄VO₃
MDL Number:MFCD00011430