Orders LinkContactUsLinkComponent
Micro Surgical Pack
Micro Surgical Pack
Micro surgical pack
Catalog # 77662-578
Supplier #:  501315
Micro Surgical Pack
Catalog # 77662-578
Micro Surgical Pack
Micro surgical pack
Price
$879.26
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Micro-surgical kit is a comprehensive kit specifically designed for microsurgical procedures.
  • No.of.piece surgical tool kit: 11
  • No.of.surgical instruments: 10
  • Autoclable instrument tray: 1
 : Kit includes: Tweezers, Dumont #5, 11 cm non-magnetic stainless steel; Scissors, spring, 14 cm curved; Needle holder, castroviejo 14 cm; Scissors, tenotomy, 10 cm straight; Instrument tray, autoclavable; Forceps, micro hemostatic, 12.5 cm, curved; Knife handle, round knurled; Blades, miniature edge 12-pack; Probe, 14 cm, stainless steel (not pictured); Retractor, weitlaner, 10.8 cm.
Specifications
  • Description:
    Micro surgical pack