ContactUsLinkComponent
Ear Tag Applicator
  77663-246
New Product
Ear Tag Applicator
  77663-246
 :  503292
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Ear tag applicator
  • Environmentally Preferable:
  • Cat. No.:
    77663-246
The ear tag in the applicator keeping the hole side of the tag flat against the jaw of the applicator with the indentation. Locate the tag within the ear, applying beyond a ring of cartilage.
After sealing, be sure to check that the tag point has come through the hole and bent around sealing properly. Tags should be washed in alcohol or similar cleaning solution to help prevent infection to the ear.
Stainless Steel Applicator is designed to seal the tags with minimal pressure, while providing a good, secure seal. Applicator can be used with or without the spring. The spring applies slight closing pressure to help hold the tag in the applicator prior to sealing.