ContactUsLinkComponent
Electrosurgical Unit, 110 V
Electrosurgical Unit, 110 V
Catalog # 77662-554
New Product
Electrosurgical Unit, 110 V
Catalog # 77662-554
Supplier Number:  501274
New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Electrosurgical unit
  • Cat. No.:
    77662-554
Specifications
About this item
Electrosurgery utilizes alternating current at radio frequencies to cut and coagulate. Using this method, the current enters the subject's body and the subject becomes part of the electrical circuit. This requires the use of a return, or ground, plate.
  • Ten levels of output intensity
  • Three operational modes (cut, coagulate, cut/coagulate)
  • Power: 110 V
This economically priced electrosurgical unit has 10 levels of output intensity, three operational modes (cut, coagulate, and cut/coagulate) and various choices of electrodes. The unit comes complete and ready-to-use with a handpiece, ground plate, foot switch and one of each electrodes.
Benefits: Ready to use out of the box, includes a variety of electrodes, which are also sold individually.
Application: Cutting and coagulation of tissue
Ordering information: All accessories can also be ordered separately.
Ball electrode
Product Information