Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IL-21 (CTGrade GMP) (from E. coli)
Human Recombinant IL-21 (CTGrade GMP) (from E. coli)
New Product
Certificates
Human Recombinant IL-21 (CTGrade GMP) (from E. coli)
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    At or below −20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.05
  • Biological Activity:
    Bioactivity assay: Proliferation of B9 cell. ED50 ≤5 ng/ml. bioactivity ≥WHO standard not available
  • Gene ID:
    Q9HBE4
  • Reconstitution Instructions:
    Sterile water at 0.1 - 0.5 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein/Peptide Name:
    CTGrade GMP IL-21
  • Purity:
    ≥97%
  • Molecular Weight:
    15.4 kDa
  • Sequence:
    MQDRHMIRMR QLIDIVDQLK NYVNDLVPEF LPAPEDVETN CEWSAFSCFQ KAQLKSANTG NNERIINVSI KKLKRKPPST NAGRRQKHRL TCPSCDSYEK KPPKEFLERF KSLLQKMIHQ HLSSRTHGSE DS
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    GMP
  • Shipping Temperature:
    Ambient
  • Tested Applications:
    Bioassay

Specifications

Frequently Bought Together