Mouse Carotid Artery Catheters, 32 G
New Product
Catheters used for sampling or dosing can vary and depend on access or animal size.
- Mouse artery catheter
- 32 Guage
- One bead
- Polyurethane
- Dead volume 37.85 microliters
All catheters are ETO sterilized.
: One per pack.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.