Human/Mouse Recombinant TGF-B 3 (from E. coli)
Supplier: Fujifilm Irvine Scientific
New Product
Certificates
About this item
Transforming growth factors (TGFs) are multifunctional peptides that regulate growth and differentiation in most cell types. The TGF-β family of proteins signal through serine/threonine kinase receptors. TGF-β isoforms (TGF-β1, -β2, and –β3) have overlapping, yet distinct biological actions in developing and adult tissues. TGF-β3 is an important factor in regulating cell adhesion and accelerating wound repair. TGF-β3 also functions during osteoblast proliferation, chemotaxis, and collagen synthesis.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:HumanMouse
- Storage Conditions:At or below –20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity Assay: Inhibition of IL-4-induced HT-2 cell proliferation, ED₅₀ ≤1 ng/ml, Bioactivity ≥1.0×10⁶ units/mg
- Gene ID:P10600/P17125
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein/Peptide Name:H/M TGF-B 3
- Purity:≥95%
- Molecular Weight:12.9/25.7 kDa
- Sequence:MALDTNYCFR NLEENCCVRP LYIDFRQDLG WKWVHEPKGY YANFCSGPCP YLRSADTTHS TVLGLYNTLN PEASASPCCV PQDLEPLTIL YYVGRTPKVE QLSNMVVKSC KCS
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay
Specifications
Frequently Bought Together