Order Entry
Puerto Rico
ContactUsLinkComponent
Human/Mouse/Rat Recombinant Activin-A (Animal free) (from E. coli)
Human/Mouse/Rat Recombinant Activin-A (Animal free) (from E. coli)
New Product
Certificates
Human/Mouse/Rat Recombinant Activin-A (Animal free) (from E. coli)
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
    Rat
    Mouse
  • Storage Conditions:
    At or below –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: Cytotoxicity of MPC-11 cells, ED50 ≤10 ng/ml, Bioactivity ≥1.0×10⁵ units/mg
  • Gene ID:
    P08476 / Q04998 / P18331
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Inhibin beta-1|FRP|FSH-releasing protein
  • Protein/Peptide Name:
    Activin-A
  • Purity:
    ≥95%
  • Molecular Weight:
    13.1/26.2 kDa
  • Sequence:
    MGLECDGKVN ICCKKQFFVS FKDIGWNDWI IAPSGYHANY CEGECPSHIA GTSGSSLSFH STVINHYRMR GHSPFANLKS CCVPTKLRPM SMLYYDDGQN IIKKDIQNMI VEECGCS
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications

Frequently Bought Together