About this item
IL-21 is a common chain cytokine regulating many cell types of the immune system. The IL-21 receptor (IL-21R) activates the JAK/STAT signaling pathway and is expressed on T, B, and natural killer (NK) cells. Plays an important role in the development of humoral immunity through its effects on B cell biology including differentiation, affinity maturation, and memory responses.
- High quality
- Low endotoxin
- Lot-to-lot consistency
- Animal free
- Manufactured following GMP guidelines
Regulates T cell responses including survival of CD8+ T cells and inhibition of Treg cells. IL-21 plays a pivotal role in Hodgkin’s lymphoma and suppresses contact hypersensitivity reactions. Promotes a less differentiated phenotype of CAR T cells while promoting the effector functions of the engineered cells. The biological activity of CTGrade GMP interleukins and growth factors is standardized, where applicable, to WHO International standards, providing cell and gene therapy developers consistent, lot-to-lot biological activity and performance.
Host cell protein: ≤30 ng/ml
Host cell DNA: ≤0.01 ng/ml
Sterility testing: No growth
Mycoplasma: Negative
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:At or below −20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.05
- Biological Activity:Bioactivity assay: Proliferation of B9 cell. ED50 ≤0.025 ng/ml. bioactivity ≥1.2 x10⁶ IU/µg
- Gene ID:P05231
- Reconstitution Instructions:Sterile water at 0.1 - 0.5 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:IFN-β2|HSF|BSF-2|B-cell Differentiation Factor|MGI-2
- UniProtKB:P05231
- Protein/Peptide Name:CTGrade GMP IL-6
- Purity:≥97%
- Molecular Weight:20.9 kDa
- Sequence:MPVPPGEDSK DVAAPHRQPL TSSERIDKQI RYILDGISAL RKETCNKSNM CESSKEALAE NNLNLPKMAE KDGCFQSGFN EETCLVKIIT GLLEFEVYLE YLQNRFESSE EQARAVQMST KVLIQFLQKK AKNLDAITTP DPTTNASLLT KLQAQNQWLQ DMTTHLILRS FKEFLQSSLR ALRQM
- Endotoxin Level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:GMP
- Shipping Temperature:Ambient
- Tested Applications:Bioassay