Order Entry
Puerto Rico
ContactUsLinkComponent
Horse Recombinant RE VEGF-165 (Animal free) (from E. coli)
Horse Recombinant RE VEGF-165 (Animal free) (from E. coli)
New Product
Certificates
Horse Recombinant RE VEGF-165 (Animal free) (from E. coli)
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Horse
  • Storage Conditions:
    −20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: HUVEC Proliferation. ED50 ≤10 ng/ml. bioactivity ≥1.0×10⁵ units/mg
  • Gene ID:
    Q9GKR0
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    glioma-derived endothelial cell mitogen|VEGF-A|VPF
  • Accession No.:
    Q9GKR0
  • Protein/Peptide Name:
    RE VEGF-165 (AF)
  • Purity:
    ≥95%
  • Molecular Weight:
    19.3/38.6 kDa
  • Sequence:
    MAPMAEGEHK THEVVKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCGGC
    CNDEGLECVP TAEFNITMQI MRIKPHQSQH IGEMSFLQHS KCECRPKKDK ARQENPCGPC
    SERRKHLFVQ DPQTCKCSCK NTDSRCKARQ LELNERTCRC DKPRR
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications

Frequently Bought Together