ContactUsLinkComponent
Needle Holder, Round Hollow Handled
Needle Holder, Round Hollow Handled
Catalog # 77661-824
New Product
Needle Holder, Round Hollow Handled
Catalog # 77661-824
Supplier Number:  14206
New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Needle holder, curved
  • Length:
    14 cm
  • Cat. No.:
    77661-824
Specifications
About this item
These round-handled needle holders are a popular choice for researchers doing small animal surgeries. The knurled handle provides a secure grip on the surgical instrument and reduces the chance of it slipping during use. The round handle is also more comfortable, especially for long procedures.
  • Smooth jaws
  • Spring handles for ambidextrous use
  • Round, hollow handles for lighter weight and firm control
  • Surgical grade stainless steel
The round handles allow you to roll the instrument between your fingers as you are working, giving you more precise control over the maneuvering of the tip holding the surgical needle. The knurled handle let you apply the correct amount of pressure for accurate suturing. The handle is hollow, reducing the weight of the instrument to also reduce hand fatigue.
The smooth jaw of the needle holder reduces the amount of trauma and tearing of delicate tissues and improves the visibility of the working area during a procedure. The tips are fine (0.3 mm) for securely grasping tiny needles. These needle holders are available in surgical grade stainless steel. Our knurled, round-handled needle holders are easy to sterilize, using a chemical sterilant or an autoclave. Always clean your surgical instruments promptly after use. This minimizes the chance of corrosion or staining and improves the longevity of your tools.