Human Recombinant MYOSTATIN (Animal free) (from E. coli)
New Product
B cell-activating factor (BAFF), or B lymphocyte stimulator (BLyS), is a type II member of the tumor necrosis factor (TNF) superfamily. BAFF is expressed as a transmembrane protein on T cells, macrophages, and dendritic cells. The transmembrane domain of BAFF can also be cleaved to produce a soluble protein fragment.
- High quality
- Low endotoxin
- Affordable
BAFF binds to the TNF receptors known as B cell maturation antigen (BCMA), transmembrane activator and CAML interactor (TACI), and BAFF receptor (BAFFR). BAFF is important for the survival and maturation of peripheral B cells. Human BAFF shows activity on mouse splenocytes.
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:−20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: MPC-11 cell cytotoxicity, ED50 ≤50 ng/ml, bioactivity ≥2.0×10⁴ units/mg
- Gene ID:O14793
- Reconstitution Instructions:Sterile 20 mM HCl at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:GDF-8|MSTN
- Accession No.:O14793
- Protein/Peptide Name:MYOSTATIN (AF)
- Purity:≥95%
- Molecular Weight:12.4/24.8 kDa
- Sequence:DFGLDCDEHS TESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGECEFV FLQKYPHTHL VHQANPRGSA GPCCTPTKMS PINMLYFNGK EQIIYGKIPA MVVDRCGCS
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay