TCLP Volatiles Spike
Catalog # SXTCLP-VX
Supplier: SPEX CERTIPREP LLC
New Product
CAS Number:
Specifications
- Cat. No.:SXTCLP-VX
- Storage temperature:Freezer
Specifications
About this item
TCLP Series Volatiles Toxicity Characteristic Leaching Procedure (TCLP) is a soil sample extraction method for chemical analysis employed as an analytical method to simulate leaching through a landfill.