Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant M-CSF (CTGrade GMP) (from E. coli)
Human Recombinant M-CSF (CTGrade GMP) (from E. coli)
New Product
Certificates
Human Recombinant M-CSF (CTGrade GMP) (from E. coli)
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    At or below –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.01
  • Biological Activity:
    Bioactivity Assay: Proliferation of NFS-60 cell, ED₅₀ ≤10 ng/ml, Bioactivity ≥1.02×10⁵  IU/µg
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    CSF-1|mgI-IM
  • Accession No.:
    P09603
  • Protein/Peptide Name:
    CTGrade GMP M-CSF
  • Purity:
    ≥97%
  • Molecular Weight:
    18.5/37.1 kDa
  • Sequence:
    MEEVSEYCSH MIGSGHLQSL QRLIDSQMET SCQITFEFVD QEQLKDPVCY LKKAFLLVQD IMEDTMRFRD NTPNAIAIVQ LQELSLRLKS CFTKDYEEHD KACVRTFYET PLQLLEKVKN VFNETKNLLD KDWNIFSKNC NNSFAECSSQ GHERQSEGS
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM Sodium phosphate and 50 mM Sodium chloride, pH 8.0
  • Nuclease-free:
    N
  • Grade:
    GMP
  • Shipping Temperature:
    Ambient
  • Tested Applications:
    Bioassay

Specifications