Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Deionized Water

Deionized Water

Deionized water, 25/pack, 5 ml
Catalog # 77623-686
Supplier:   Hardy Diagnostics
Supplier #:  X58
Deionized Water
Catalog # 77623-686
Supplier:  Hardy Diagnostics
CAS Number:  
Deionized Water

Deionized water, 25/pack, 5 ml

CAS Number:  
Price
$113.00
Log in for availability
Pack of 25 (5ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

For the preparation of sample dilutions.

  • Sterilized by autoclaving
  • Supplied in Centrifuge tube
Specifications

Specifications

  • Pack type:
    25/pack
  • Description:
    Deionized water
  • Fill volume:
    5 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.