Order Entry
United States
ContactUsLinkComponent
 
Wecker Spatula
  77662-836
New Product
Wecker Spatula
  77662-836
 :  501772
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Length:
    14 cm (5.5")
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Cat. No.:
    77662-836
  • Blade width:
    3 mm
  • Blade type:
    Flat

 

 

A Wecker Spatula is a surgical instrument designed for ophthalmology to lift, separate, dissect or manipulate delicate tissues of the eye like the conjunctiva or cornea.

  • Thin, flat blade with rounded tip
  • 3 mm wide blade
  • Designed for ophthalmology, great for microdissection

This Wecker spatula has a fine, narrow blade. It has a solid square handle for a secure grip. This spatula can be sterilized with a chemical sterilant or by autoclaving. Always clean the surgical instruments promptly after use to ensure they last a long time.

The design of the Wecker Spatula allows for precise manipulation and dissection. In a research laboratory, the fine blade of the Wecker spatula makes it ideal for microdissection, which involves the precise isolation and removal of small tissue samples or specific structures under a microscope.

Its fine blade allows for precise cutting and manipulation without causing significant damage to surrounding tissues. Or, the Wecker Spatula can be used to isolate individual cells or small clusters of cells from a heterogeneous population. By gently scraping or lifting targeted cells, researchers can separate them for further analysis. Or, you could use the Wecker Spatula to remove unwanted debris, contaminants, or non-specific tissue components from a sample.