Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant FGF-21 (Animal Free) (from E. coli)
Human Recombinant FGF-21 (Animal Free) (from E. coli)
New Product
Certificates
Human Recombinant FGF-21 (Animal Free) (from E. coli)
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity assay: NIH-3T3 proliferation with β-klotho and heparin, ED50 ≤500 ng/ml, Bioactivity ≥2.0 ×10³ unit/mg
  • Gene ID:
    Q9NSA1
  • Amino Acid Number:
    182
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    FGFL
  • Protein/Peptide Name:
    FGF-21
  • Purity:
    ≥95%
  • Molecular Weight:
    19.5 kDa
  • Sequence:
    MHPIPDSSPL LQFGGQVRQR YLYTDDAQQT EAHLEIREDG TVGGAADQSP ESLLQLKALK PGVIQILGVK TSRFLCQRPD GALYGSLHFD PEACSFRELL LEDGYNVYQS EAHGLPLHLP GNKSPHRDPA PRGPARFLPL PGLPPALPEP PGILAPQPPD VGSSDPLSMV GPSQGRSPSY AS
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM sodium phosphate, 100 mM sodium chloride, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

Specifications