Orders LinkContactUsLinkComponent
Rat Kit for Telemetry Training
Rat Kit for Telemetry Training
Basic rat kit for Telemetry training (8 piece)
Catalog # 77663-754
Supplier #:  504740
Rat Kit for Telemetry Training
Catalog # 77663-754
Rat Kit for Telemetry Training
Basic rat kit for Telemetry training (8 piece)
Price
$605.18
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Basic 8 piece kit for performing rat surgeries and dissections.
 : Kit includes: (1) Dumont #5 tweezers; (1) Alm self retaining retractor; (1) Vannas scissors; (1) Needle holder; (1) Iris forceps, curved, serrated; (1) Blunt probe, 1.0 mm diameter; (1) SuperCut Iris scissors, straight, 10 cm; (1) Forceps vessel cannulation.
Specifications
  • Description:
    Basic rat kit for Telemetry training (8 piece)