Mouse Recombinant IL-21 (Animal free) (from E. coli)
New Product
Interleukin 21 (IL-21) is a member of the common-gamma chain family of cytokines that are involved in immunoregulation. IL-21 is normally expressed by activated CD4+ T cells. The IL-21 receptor (IL-21R) activates the JAK/STAT signaling pathway and is expressed on T, B, and natural killer (NK) cells. Within the B cell lineage, IL-21 is a switch factor regulating IgG1 and IgG3 antibody production. IL-21 also cooperates with interleukin 4 (IL-4) for the production of multiple antibody classes in B cells. IL-21 has pleiotropic effects on the proliferation, differentiation, and effector functions of B, T, NK, and dendritic cells.
- High Quality
- Low Endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: proliferation of B9 cells, ED50 ≤20 ng/ml, Bioactivity ≥1.0×10⁵ units/mg or 5.0×10⁴ units/mg
- Gene ID:Q9ES17
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein/Peptide Name:IL-21
- Purity:≥95%
- Molecular Weight:15.1 kDa
- Sequence:MHKSSPQGPD RLLIRLRHLI DIVEQLKIYE NDLDPELLSA PQDVKGHCEH AAFACFQKAK LKPSNPGNNK TFIIDLVAQL RRRLPARRGG KKQKHIAKCP SCDSYEKRTP KEFLERLKWL LQKMIHQHLS
- Endotoxin Level:Low
- Formulation:20 mM sodium bicarbonate, pH 8.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay
Frequently Bought Together