Orders LinkContactUsLinkComponent
Iris Scissors
Iris Scissors
Iris Scissors
Certificates
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications.
  • Ring finger handle
  • Stainless steel
The tips are sharp, and they come in curved, straight or angled styles.
Product Family Options
Product Information
  • Length
    Material
    Blade length
    Availability
    Price
  • 77662-816
    Iris scissor, straight tips
    Iris scissor, straight tips
    11.5 cm (4.5")
    Stainless steel
    2.2 cm
    Log in for availability
    Log in for availability
    Each
    Each - $73.94
    $73.94
    Each
       $73.94
    Supplier Number: 501758
  • 77662-818
    Iris scissor, straight tips
    Iris scissor, straight tips
    11.5 cm (4.5")
    German steel
    2.2 cm
    Log in for availability
    Log in for availability
    Each
    Each - $54.00
    $54.00
    Each
       $54.00
    Supplier Number: 501758-G
  • 77662-820
    Iris scissor, curved tips
    Iris scissor, curved tips
    11.5 cm (4.5")
    Stainless steel
    2.2 cm
    Log in for availability
    Log in for availability
    Each
    Each - $73.94
    $73.94
    Each
       $73.94
    Supplier Number: 501759
  • 77662-822
    Iris scissor, curved tips
    Iris scissor, curved tips
    11.5 cm (4.5")
    German steel
    2.2 cm
    Log in for availability
    Log in for availability
    Each
    Each - $54.00
    $54.00
    Each
       $54.00
    Supplier Number: 501759-G
  • 77663-156
    Iris scissor, 35° angled, serrated tips
    Iris scissor, 35° angled, serrated tips
    11.5 cm (4.5")
    Stainless steel
    2.2 cm
    Log in for availability
    Log in for availability
    Each
    Each - $113.96
    $113.96
    Each
       $113.96
    Supplier Number: 503224
  • 77663-158
    Iris scissor, 45° angled, serrated tips
    Iris scissor, 45° angled, serrated tips
    11.5 cm (4.5")
    Stainless steel
    2.2 cm
    Log in for availability
    Log in for availability
    Each
    Each - $120.13
    $120.13
    Each
       $120.13
    Supplier Number: 503225