Racks for Ivaro Device
Catalog # 77589-636
Supplier: SARSTEDT INC
New Product
Specifications
- Color:White
- Description:Rack for ivaro
- Material:POM
- Cat. No.:77589-636
- Packaging:Blister
Specifications
About this item
Accessories to support your workflow with the IVARO device.
- Fully automated uncapping and recapping, filling and labeling
- Individual device configuration for application and long stand-alone times
- Highly flexible as a wide range of tubes can be processed
IVARO FD - Optimal filling and dosing the IVARO FD is ideal solution when small containers need to be filled with stock solutions, buffers and other liquids. Preparing samples, producing small product series, manufacturing kits and other time-intensive applications are made easy with the IVARO.
IVARO AP - Precise aliquoting and pipetting. The IVARO AP is more than a liquid handler. It supports you in all types of sample preparation. It can easily fill several identical tubes simultaneously with liquid and aliquot specimens into different containers.