Sealing Clips
Catalog # 77589-604
Supplier: SARSTEDT INC
New Product
Specifications
- Description:Sealings clip
- Cat. No.:77589-604
- Packaging:BAGA
Specifications
About this item
SARSTEDT transfusion products are user-friendly, efficient and safe.
- Stripper/sealer provides easy handling thanks to the ergonomically designed handle
- Made of aluminium
- Adjustable to different tube diameters
For manual stripping and sealing of blood bags.