Round Cuvette
Round cuvette, BAGA
Catalog # 77588-384
Supplier: SARSTEDT INC
Supplier #: 68.752
Jump to:
Product Details & Documents
SARSTEDT has been known as a manufacturer of high-quality disposable cuvettes.
- Height: 51 mm
- Material: PP
- Made of high-quality polypropylene
- Especially for specimen preparation
- Suitable for LKB analyzer (enzyme analyzer)
SARSTEDT cuvettes are made from polystyrene (PS), acrylic (PMMA) and a special plastic for analysis in the UV range, which are ideal for photometric analysis of liquid solutions. The cuvettes are also available in 2× optical semi-micro and a 4× optical format for 90° angle fluorescence measurements.
Packaging: Packaged 1000 pieces per bag.
Specifications
- Description:Round cuvette
- Packaging:BAGA



