Cell Culture Chambers, x-well Cover Glass
New Product
To keep cells happy, the selection of a suitable cultivation surface is essential. SARSTEDT offers a wide range of cell culture labware and accessories for many applications.
- Design: On cover glass
- Removable frame: No
- Cell type: Adherent cells
- Material: Borosilicate glass
- Frame material: PS
- Purity: Sterile, non-pyrogenic/endotoxin-free, non-cytotoxic
- Cell culture chambers for the analysis of adherent cell growth on cover glass
The x-well cell culture system is available in various slide material options for a multitude of microscopic cell culture analyses. PCA, glass, coverglass or lumox® film slides combined with a polystyrene frame form one- and multiple-chamber vessels. x-well cell culture chambers make parallel tests quick and easy.