Multiwell Plates, lumox® multiwell
Supplier: SARSTEDT INC
New Product
Certificates
About this item
To keep cells happy, the selection of a suitable cultivation surface is essential. SARSTEDT offers a wide range of cell culture labware and accessories for many applications.
- Cell type: Adherent cells
- Material: lumox®
- Frame material: PS
- Purity: Sterile, non-pyrogenic/endotoxin-free, non-cytotoxic
- Multi-well plates with lumox® film base and black polystyrene body/frame
- Free standing wells with alphanumeric labels minimize the risk of contamination during pipetting
- Low autofluorescence and higher light transmission than polystyrene or glass bases
- Gas-permeable film base guarantees optimal gas exchange
- Range of applications from normal cell culture to the automated analysis of fluorescence-based cell assays
The thin, gas-permeable film base of lumox® cell products ensures optimal cell growth through short diffusion paths and excellent gas exchange. lumox® film bases also have excellent optical properties. Minimal autofluorescence and high light transmission result in a high sensitivity with fluorescence-based assays. SARSTEDT offers lumox® dishes and plates as well as x-well cell culture chambers for direct cultivation and analysis of cells on a slide.
Frequently Bought Together