Orders LinkContactUsLinkComponent
Westcott Student Spring Scissors
Westcott Student Spring Scissors
Westcott Student Spring Scissors
Westcott student spring scissors are popular spring handled scissors that automatically reopen after each cut. Westcott brand spring scissors give you an ergonomic and comfortable grip, reducing hand strain and fatigue during repetitive cutting tasks.
  • Slightly curved tip gives you greater visibility at the incision site
Westcott spring scissors are perfect for ambidextrous use, often preferred by left-handed individuals. The tips of these surgical scissors are slightly curved, and they are available with blunt or sharp tips. These Westcott scissors are perfect for student use. They are constructed of surgical grade stainless steel and are suitable for autoclaving or chemical sterilization.
Product Family Options
  • Description
    Length
    Material
    Blade type
    Curvature
  • Wescott student scissors
    11 cm (4.3")
    Stainless steel
    Blunt
    Slightly curved
    Each
    Each - $174.02
    $174.02
    Each
       $174.02
     : 503222
  • Wescott student scissors
    11 cm (4.3")
    Stainless steel
    Sharp
    Slightly curved
    Each
    Each - $174.02
    $174.02
    Each
       $174.02
     : 503223