Inoculation Spreader
Inoculation spreader, Blue, Blister pack
Catalog # 77589-582
Supplier: SARSTEDT INC
Supplier #: 86.1569.005
Jump to:
Product Details & Documents
One aspect of microbiology is researching and identifying pathogens, as well as looking for suitable treatments. SARSTEDT provides researchers with the consumables they need to do their work.
- Material: PS
- Ideal for evenly spreading suspensions with pathogens on the surface of agar plates (e.g. for determining the number of live cells)
- No damage to the culture medium thanks to the smooth and even surface
- Saves time when processing larger test series
- Safe handling - no pathogenic aerosols or cross contamination
- Sterilized by gamma irradiation
Flexible SARSTEDT inoculation loops allow swabs to be taken easily or inoculation in liquid media. The inoculation needle can be used to inoculate or remove individual colonies. We recommend use of the inoculation spreader to evenly apply larger volumes to culture media.
Specifications
- Color:Blue
- Description:Inoculation spreader
- Packaging:Blister pack