Conical Centrifuge Tubes
New Product
High-quality, safe and robust containers are essential for mixing, processing and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
- Material: PP
- Cap material: HD-PE
- Graduations: Yes
- Blue graduated scale for ideal volume reading
- Resilient text field allows clear labelling
- Certified free of DNA, DNase, RNase, PCR inhibitors, pyrogens and non-cytotoxic
- Especially leak tight as per the ADR transport directive
This broad product portfolio in the field of reagent and centrifuge tubes includes options with round, conical or flat base; with or without skirted base; with stopper or screw cap; with or without print or labels.