ContactUsLinkComponent
Digital Microscopes for Inspection, Emspira 3, Leica Microsystems
Digital Microscopes for Inspection, Emspira 3, Leica Microsystems
Catalog # 77676-434
Supplier:  Leica Microsystems
New Product
undefined
Digital Microscopes for Inspection, Emspira 3, Leica Microsystems
Catalog # 77676-434
Supplier:  Leica Microsystems
Supplier Number:  8122685
New Product
Specifications
  • Description:
    Leica Emspira 3 digital microscopes for inspection
  • Cat. No.:
    77676-434
Specifications
About this item
The Leica Microsystems Emspira 3 digital microscope is a cutting-edge tool for precision inspection. With advanced digital capabilities and high-quality imaging, it offers detailed visualisations. Elevate your inspection workflow with this user-friendly and versatile microscope.
  • Measure and annotate directly during visual inspection without a PC
  • 8:1 zoom ratio with 26× to 206× magnification range (1× Planapo objective, optional 28" - monitor) and parfocal optics - from overview to detail
  • Minimise the risk of data loss by keeping results and data directly on your local network for worry-free sharing
  • Customisable User Interface (UI) for simplified workflows
The Leica Emspira 3 digital microscope is a cutting-edge tool tailored for precision inspection tasks. With advanced digital capabilities and high-quality imaging, this microscope transforms the inspection process, providing detailed and clear visualizations. Its user-friendly design and versatility make it a smart choice for professionals seeking efficiency and accuracy in inspection applications. Elevate your inspection workflow with the Leica Emspira 3, a digital microscope designed to meet the demands of meticulous inspection tasks with exceptional clarity and ease.
Features and specifications include:
Stand alone operation
Control via mouse, keyboard, foot/hand switch and/or touchscreen
Integrated measurement, annotation and sharing functionality
Create overlays and reference images for comparison
Capture directly to USB stick or network folder
Send images via e-mail
4k 60fps live image via HDMI
Antimicrobial PC/ABS housing with AgTreat™
Possible magnification range 8.2× to 1027× (depending on objective with 28" monitor)
Coded zoom with click stop option for reproducible workflows
JPG/TIFF/BMP images with up to 12 MP
Full HD MJPG Video recording with 30 fps
USB 3.1 type C connection
Control via enersight mobile (iOS/android) or via enersight OSD
Delivery information: Includes: Emspira 3; samsung 4k HDMI monitor; power cable, 2 m, USA; WIFI dongle 5GHZ für FLEXACAM C1 (US); 1D barcodescanner; focus drive 500 mm M-series routine; intermediate adapter for TL and IL base; microscope carrier; inc. light base, large w. antishock feet; Leica LED3000 RL, 58mm; polarisation set for LED3000 RL; dust cover antistatic, small; objective 1.0× planapo Z-series; adapter-telecentric, 0.8× and 1.0× planapo; standalone kit for Emspira 3.