- Description:Leica Ivesta 3 Stereo microscopes for inspection with integrated camera
- Accessories Included:HDMI cable, USB C-A cable, external power supply (without power cord), USB stick
- Cat. No.:77676-430
The Leica Ivesta 3 stereo microscope offers Leica optics and advanced imaging for inspection and rework. Its integrated camera enables efficient documentation and analysis. Elevate work processes with this reliable microscope, enhancing accuracy and productivity in inspection and rework tasks.
- Change from sample overview to detail easily with the microscope's large 9:1 zoom range
- 122 mm standard working distance (without lens shield) - ensures excellent accessibility to the samples
- Save time refocusing as you can see every important detail at a glance
- FusionOptics allows you to perceive your samples in 3D with up to 3× larger depth of field
- Reveal fine details with a magnification of up to 55× and apochromatically corrected optics.
- Enable fast and easy discussions of results to get other opinions: Fast display of live images from the 12-megapixel camera on a monitor
The Leica Ivesta 3 stereo microscope is a powerhouse for inspection and rework, featuring an integrated camera for documentation. Designed for professionals, this microscope combines Leica optics with advanced imaging capabilities, providing unparalleled clarity in inspection and rework tasks. The integrated camera adds an extra layer of efficiency, allowing users to document and analyze their work seamlessly. Elevate your work processes with the Leica Ivesta 3, a reliable stereo microscope that seamlessly integrates cutting-edge technology to enhance accuracy and productivity in inspection and rework applications.
Features and specifications include:
9:1 zoom with magnification 6.1 to 55× (10× eyepieces, no supplementary lens) - provides field of view up to 37.6 mm for fast, accurate inspection
12 mm depth of field with fusion optics. Greenough optical design - provides very flat (plan) field of view and large depth of field
Complete apochromatic corrected microscopy system - 35° angle eye tubes - for comfort and convenience
122 mm standard working distance (without lens shield) - ensures excellent accessibility to the samples
Resolution up to 250 lp/mm (without supplementary lens, 10× eyepieces), maximum resolution up to 500 lp/mm (2× supplementary lens, 10× eyepieces)
Fully encoded zoom - inspection at specific magnification can be easily repeated
Antimicrobial PC/ABS housing with AgTreat™
Large range of supplementary lenses as well as eyepieces
Adapt the optical performance to the application requirements
Integrated digital color camera with CMOS sensor
Possible to operate with 4k HDMI monitor using enersight OSD
High definition live image 4k, 60fps, HDR
JPG images with up to 12 MP
Camera control via Windows using Enersight desktop or via tablet/smartphone via enersight mobile application
Capture images directly to USB stick
USB-C connection to PC for enersight desktop