ContactUsLinkComponent
Stereo Microscope for Routine Sorting, Ivesta 3, Leica Microsystems
Stereo Microscope for Routine Sorting, Ivesta 3, Leica Microsystems
Catalog # 77676-398
Supplier:  Leica Microsystems
New Product
undefined
Stereo Microscope for Routine Sorting, Ivesta 3, Leica Microsystems
Catalog # 77676-398
Supplier:  Leica Microsystems
Supplier Number:  8122664
New Product
Specifications
  • Description:
    Leica Ivesta 3 for routine sorting
  • Cat. No.:
    77676-398
Specifications
About this item
The Leica Ivesta 3 stereo microscope offers precise optics and user-friendly features, enhancing sorting tasks in labs. With advanced capabilities, it streamlines processes, providing reliable and efficient results. Elevate your workflow with the convenience of the Leica Ivesta 3 microscope.
  • Change from sample overview to detail easily with the microscope's large 9:1 zoom range
  • 122 mm standard working distance (without lens shield) - ensures excellent accessibility to the samples
  • Save time refocusing as you can see every important detail at a glance
  • FusionOptics allows you to perceive your samples in 3D with up to 3× larger depth of field
  • Reveal fine details with a magnification of up to 55× and apochromatically corrected optics
The Leica Ivesta 3 stereo microscope is for routine sorting applications. Engineered with precision optics and user-friendly features, this microscope offers exceptional clarity and ease of use. With its advanced capabilities, the Leica Ivesta 3 streamlines routine cell sorting processes, providing researchers and professionals with a reliable and efficient tool for daily laboratory tasks. Elevate your workflow with the performance and convenience offered by the Leica Ivesta 3 stereo microscope.
Features and specifications include:
9:1 zoom with magnification 6.1 to 55× (10× eyepieces, no supplementary lens) - provides field of view up to 37.6 mm for fast, accurate sorting
12 mm depth of field with fusion optics. Greenough optical design - provides very flat (plan) field of view and large depth of field
Complete apochromatic corrected microscopy system - 35° angle eye tubes - for comfort and convenience
122 mm standard working distance (without lens shield) - ensures excellent accessibility to the samples
Resolution up to 250 lp/mm (without supplementary lens, 10× eyepieces), maximum resolution up to 500 lp/mm (2× supplementary lens, 10× eyepieces)
Antimicrobial PC/ABS housing with AgTreat™
Clickstops at 10×, 20×, 30×, 40× and 50× - inspection at specific magnification can be easily repeated
Large range of supplementary lenses as well as eyepieces
Delivery: Includes: Ivesta 3; eyepiece 10×/23B, adj., eyeglasses x²; focus column for S-line; incident light base, small; sub base with reflector; KL300 LED fiberoptic light source w/ps; KL300 LED bracket for routine focusdrive; universal light guide, 600 mm; dust cover antistatic, small.