ContactUsLinkComponent
Inverted Microscope for Core Cell Culture, DMi1, Leica Microsystems
Inverted Microscope for Core Cell Culture, DMi1, Leica Microsystems
Catalog # 77676-390
Supplier:  Leica Microsystems
New Product
undefined
Inverted Microscope for Core Cell Culture, DMi1, Leica Microsystems
Catalog # 77676-390
Supplier:  Leica Microsystems
Supplier Number:  8122660
New Product
Specifications
  • Description:
    Leica DMi1 for core cell culture
  • Accessories Included:
    Dust cover
  • Condenser:
    S80
  • Cat. No.:
    77676-390
Specifications
About this item
The Leica DMi1 is a high-quality manual inverted microscope, perfect for cell culture labs. It offers exceptional clarity, precision optics, and user-friendly features, making it designed for observing cell cultures and individual cells. Elevate your research with its reliable imaging capabilities.
  • Easy-to-use operation that requires minimal training and maintenance
  • Cool, color-safe LED illumination for constant color temperature
  • Easy phase contrast: 10×, 20×, and 40× (not included) objectives share the same light ring
  • World class Leica objectives that provide brilliant images
The Leica DMi1 for Cell Culture is a robust manual inverted microscope designed for routine tasks in cell culture labs. With precision optics and user-friendly features, this microscope delivers exceptional clarity and detail, making it a tool designed for observing cell cultures and individual cells. Elevate your cell culture research with the Leica DMI1's outstanding performance and reliable imaging capabilities.
Features and specifications include:
Leica DMi1 FOV20 basic stand FOV20, exchangeable eyepieces, fix stage, power cable
Two eyepieces(10×/20 eyepiece w/yeguard and 10×/20 focusing eyepiece w/eyeguard)
Four position slider for S80 condenser(BF, Ph0, Ph1 and Ph2)
Three objectives(HI PLAN 4×/0.10 PH0, HI PLAN I 10×/0.22 PH1, HI PLAN I 20×/0.30 PH1)
Delivery: Includes DMi1 FOV20, S80, 4P slider, 3 obj. IVD.