Serological Pipettes, Graduated, Individually Wrapped
Supplier: SARSTEDT INC
New Product
Certificates
About this item
Dosing, transferring, and measuring liquids are routine procedures in almost every laboratory. SARSTEDT offers a wide range of products for every application, from high-quality pipette tips and serological pipettes to transfer pipettes, for reliable and reproducible results.
- Sterile, non-pyrogenic/endotoxin-free, non-cytotoxic
- Easy volume identification using the international color code on each pipette and blister packaging
- Optimized mouthpieces for universal and secure fit in common pipetting aids
- Larger pipetting volume due to negative scaling
- Variable method of work due to counter scaling
- Easy-to-open, anti-static packaging
SARSTEDT serological pipettes provide controlled measuring of larger volumes of liquid up to 50 ml. Individually wrapped versions in all sizes are available for safe and sterile work.