Serological Pipettes, Graduated
Supplier: SARSTEDT INC
New Product
Certificates
About this item
Dosing, transferring, and measuring liquids are routine procedures in almost every laboratory. SARSTEDT offers a wide range of products for every application, from high-quality pipette tips and serological pipettes to transfer pipettes, for reliable and reproducible results.
- Easy volume identification using the international color code on each pipette
- Optimized mouthpieces for universal and secure fit in common pipetting aids
- Larger pipetting volume due to negative scaling
- Variable method of work due to counter scaling
SARSTEDT serological pipettes provide controlled measuring of larger volumes of liquid up to 25 ml.