Orders LinkContactUsLinkComponent
Forensics Smear Swabs
Forensics Smear Swabs
Supplier:   SARSTEDT INC
Forensics Smear Swabs
Supplier:  SARSTEDT INC
Certificates
Forensics Smear Swabs
Forensics Smear Swabs
5 Options Available Below
Product Details & Documents
SARSTDT forensic swabs contain a ventilation membrane in the tube base, allowing the swab to dry in the tube.
  • Round swab shape
  • Complies with all requirements of ISO 18385
  • Employee DNA exclusion database available
An EtO gassing process developed specifically to address the requirements of forensics, as well as the highest cleanroom conditions in the production process, guarantee that these forensic swabs are absolutely free from DNA.
Product Family Options
Product Information
  • Handle material
    Handle length
    Head material
    Availability
    Price
  • 77589-136
    Forensics smear swab, with ventilation membrane
    Forensics smear swab, with ventilation membrane
    PS
    85 mm
    Viscose
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $142.09
    $142.09
    Log in for availability
    Case of 500
    Case of 500 - $1,420.90
    $1,420.90
    Pack of 50
       $142.09
    Case of 500
       $1,420.90
    Supplier Number: 80.630
  • 77589-132
    Forensics smear swab, with ventilation membrane
    Forensics smear swab, with ventilation membrane
    PS
    93 mm
    Viscose
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $277.13
    $277.13
    Log in for availability
    Case of 800
    Case of 800 - $2,217.04
    $2,217.04
    Pack of 100
       $277.13
    Case of 800
       $2,217.04
    Supplier Number: 80.629
  • 77589-134
    Forensics smear swab, with ventilation membrane
    Forensics smear swab, with ventilation membrane
    Wood
    95 mm
    Cotton
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $300.37
    $300.37
    Log in for availability
    Case of 800
    Case of 800 - $2,402.96
    $2,402.96
    Pack of 100
       $300.37
    Case of 800
       $2,402.96
    Supplier Number: 80.629.001
  • 77589-138
    Forensics smear swab, with ventilation membrane
    Forensics smear swab, with ventilation membrane
    PS
    125 mm
    Viscose
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $142.09
    $142.09
    Log in for availability
    Case of 500
    Case of 500 - $1,420.90
    $1,420.90
    Pack of 50
       $142.09
    Case of 500
       $1,420.90
    Supplier Number: 80.634
  • 77589-130
    Forensics smear swab, without ventilation membrane
    Forensics smear swab, without ventilation membrane
    Wood
    150 mm
    Cotton
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $49.08
    $49.08
    Log in for availability
    Case of 1000
    Case of 1000 - $490.80
    $490.80
    Pack of 100
       $49.08
    Case of 1000
       $490.80
    Supplier Number: 80.626