ContactUsLinkComponent
Wastewater inorganics quality control standard
  MSPP-2833249
 :  
undefined
Wastewater inorganics quality control standard
  MSPP-2833249
 :  2833249
 :  
  • Concentration:
    2 mg/L as NH3-N
    4 mg/L as NO3-N
    2 mg/L as PO43-
    25 mg/L as O2
    50 mg/L as SO42-
    8 mg/L as C
  • Description:
    Wastewater effluent inorganics quality control standard, 500 ml
  • Cat. No.:
    MSPP-2833249
These mixed standards are ready to use, no dilutions are required. They contain ammonia nitrogen, nitrate nitrogen, COD, phosphate, sulphate and TOC.